Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTACAATCAAAGGGAGAGGAGA[A/G]GAGTCAGTGGGGGCAGGGGAAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605461 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CECR6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CECR6 - cat eye syndrome chromosome region, candidate 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163079.1 | 3031 | UTR 3 | NP_001156551.1 | |||
NM_031890.3 | 3031 | UTR 3 | NP_114096.1 | |||
XM_011546124.2 | 3031 | Intron | XP_011544426.1 | |||
XM_011546125.2 | 3031 | UTR 3 | XP_011544427.1 |
IL17RA - interleukin 17 receptor A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100996342 - uncharacterized LOC100996342 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |