Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCCAACTGCTGTCCAGGTTCAT[A/G]GATAGTCATGCTTGGTCTTGTCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612824 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC14L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC14L3 - SEC14 like lipid binding 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257378.1 | Intron | NP_001244307.1 | ||||
NM_001257379.1 | Intron | NP_001244308.1 | ||||
NM_001257382.1 | Intron | NP_001244311.1 | ||||
NM_174975.4 | Intron | NP_777635.1 | ||||
XM_011530128.2 | Intron | XP_011528430.1 |