Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATACCAGAGATGGACAGCCATC[G/T]GGTAGAGAAGTTGGGCCAGCACCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605819 MIM: 613441 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PES1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PES1 - pescadillo ribosomal biogenesis factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCN2 - transcobalamin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000355.3 | 338 | Missense Mutation | CGG,CTG | R,L 30 | NP_000346.2 | |
NM_001184726.1 | 338 | Missense Mutation | CGG,CTG | R,L 30 | NP_001171655.1 |