Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGCACGGCTCTGGTAGGAATTG[A/C]ACAGGTTTTGGAGCCCTACTTCATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616661 | ||||||||||||||||||||
Literature Links: |
MORC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MORC2 - MORC family CW-type zinc finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303256.2 | 3283 | Missense Mutation | TGC,TTC | C,F 962 | NP_001290185.1 | |
NM_001303257.2 | 3283 | Missense Mutation | TGC,TTC | C,F 962 | NP_001290186.1 | |
NM_014941.3 | 3283 | Missense Mutation | TGC,TTC | C,F 900 | NP_055756.1 | |
XM_011530004.1 | 3283 | Missense Mutation | TGC,TTC | C,F 967 | XP_011528306.1 | |
XM_017028667.1 | 3283 | Missense Mutation | TGC,TTC | C,F 967 | XP_016884156.1 | |
XM_017028668.1 | 3283 | Missense Mutation | TGC,TTC | C,F 900 | XP_016884157.1 |
MORC2-AS1 - MORC2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |