Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCATCTGTGCCTTTATCCTGGA[A/G]CTGGTGTCGCCTCTGCTGAAGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606481 MIM: 611651 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5J PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INPP5J - inositol polyphosphate-5-phosphatase J | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLA2G3 - phospholipase A2 group III | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015715.4 | 1643 | Missense Mutation | CTC,TTC | L,F 464 | NP_056530.2 | |
XM_011530204.1 | 1643 | Missense Mutation | CTC,TTC | L,F 283 | XP_011528506.1 | |
XM_011530205.1 | 1643 | Missense Mutation | CTC,TTC | L,F 283 | XP_011528507.1 |