Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTATCATTGATGGACTTTTGGG[C/T]TGGAAGTGGGATTTAGAGATCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605969 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFPL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RFPL2 - ret finger protein like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098527.2 | Intron | NP_001091997.2 | ||||
NM_001159545.1 | Intron | NP_001153017.1 | ||||
NM_001159546.1 | Intron | NP_001153018.1 | ||||
NM_006605.3 | Intron | NP_006596.2 | ||||
XM_011529826.1 | Intron | XP_011528128.1 | ||||
XM_011529827.2 | Intron | XP_011528129.1 | ||||
XM_011529828.1 | Intron | XP_011528130.1 | ||||
XM_011529830.2 | Intron | XP_011528132.1 | ||||
XM_011529832.1 | Intron | XP_011528134.1 | ||||
XM_017028535.1 | Intron | XP_016884024.1 | ||||
XM_017028536.1 | Intron | XP_016884025.1 |