Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCCACGTGATTTCTCAGGATAA[C/T]CTGGAACAAAGATAAGCAAATGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC5A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC5A4 - solute carrier family 5 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014227.2 | 1651 | Missense Mutation | GAT,GGT | D,G 590 | NP_055042.1 | |
XM_006724308.3 | 1651 | Missense Mutation | GAT,GGT | D,G 544 | XP_006724371.1 | |
XM_011530342.2 | 1651 | Missense Mutation | GAT,GGT | D,G 544 | XP_011528644.1 | |
XM_011530343.2 | 1651 | Missense Mutation | GAT,GGT | D,G 544 | XP_011528645.1 | |
XM_011530344.2 | 1651 | Missense Mutation | GAT,GGT | D,G 521 | XP_011528646.1 | |
XM_017028920.1 | 1651 | Missense Mutation | GAT,GGT | D,G 620 | XP_016884409.1 |