Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCTTCGTCGGAGCCATCCCCGG[C/T]GCCTTTCCGGGCCATTCGGCGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160775 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6819 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6819 - microRNA 6819 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYH9 - myosin, heavy chain 9, non-muscle | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002473.5 | 5850 | Missense Mutation | ACC,GCC | T,A 1939 | NP_002464.1 | |
XM_011530197.2 | 5850 | Missense Mutation | ACC,GCC | T,A 1939 | XP_011528499.1 | |
XM_017028803.1 | 5850 | Missense Mutation | ACC,GCC | T,A 1939 | XP_016884292.1 | |
XM_017028804.1 | 5850 | Missense Mutation | ACC,GCC | T,A 1939 | XP_016884293.1 | |
XM_017028805.1 | 5850 | Missense Mutation | ACC,GCC | T,A 1939 | XP_016884294.1 | |
XM_017028806.1 | 5850 | Missense Mutation | ACC,GCC | T,A 1939 | XP_016884295.1 |