Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGTGGCAATGCTGAGAGAGTGC[C/T]GAGGAAAGAGTGGAGAGTTTGTAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615870 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFT27 - intraflagellar transport 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177701.2 | Intron | NP_001171172.1 | ||||
NM_006860.4 | Intron | NP_006851.1 | ||||
XM_006724106.2 | Intron | XP_006724169.1 | ||||
XM_017028540.1 | Intron | XP_016884029.1 |
LOC105373021 - uncharacterized LOC105373021 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |