Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTTGAGGCAACCACATCCACTT[G/T]GGAGACACGCATATGGTATGATACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615323 MIM: 607046 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JOSD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JOSD1 - Josephin domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014876.5 | 2727 | UTR 3 | NP_055691.1 | |||
XM_005261876.2 | 2727 | UTR 3 | XP_005261933.1 | |||
XM_005261877.2 | 2727 | UTR 3 | XP_005261934.1 | |||
XM_005261878.2 | 2727 | UTR 3 | XP_005261935.1 | |||
XM_005261879.2 | 2727 | UTR 3 | XP_005261936.1 |
TOMM22 - translocase of outer mitochondrial membrane 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |