Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTAATAAAGAGCAATTTTCAAG[A/G]ATATCTTTTTTGTGCTGACCATTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606795 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC25A17 - solute carrier family 25 member 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282726.1 | 1886 | UTR 3 | NP_001269655.1 | |||
NM_001282727.1 | 1886 | UTR 3 | NP_001269656.1 | |||
NM_006358.3 | 1886 | UTR 3 | NP_006349.1 |