Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGACCCGGCGAAGAGAAAAAGGA[A/G]CTTCCCCCACCCCCTCGGGTGCCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602700 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EP300 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EP300 - E1A binding protein p300 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001429.3 | 263 | UTR 5 | NP_001420.2 | |||
XM_006724165.3 | 263 | Intron | XP_006724228.1 |
MIR1281 - microRNA 1281 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |