Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATACAGGTTGCAGATAATAATTTG[C/T]TTAACTTGTGCAGTTTAATCAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RRP7BP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RRP7BP - ribosomal RNA processing 7 homolog B, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERHL2 - serine hydrolase-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284334.1 | Intron | NP_001271263.1 | ||||
NM_014509.4 | Intron | NP_055324.2 | ||||
XM_017028738.1 | Intron | XP_016884227.1 | ||||
XM_017028739.1 | Intron | XP_016884228.1 |