Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTGCATCCACCACAAGTCCGCC[A/G]GCACATCGGGCAGCCACACCACCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611589 | ||||||||||||||||||||
Literature Links: |
PNPLA5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PNPLA5 - patatin like phospholipase domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177675.1 | 900 | Missense Mutation | CCG,CTG | P,L 256 | NP_001171146.1 | |
NM_138814.3 | 900 | Missense Mutation | CCG,CTG | P,L 370 | NP_620169.1 | |
XM_011529959.2 | 900 | Missense Mutation | CCG,CTG | P,L 230 | XP_011528261.1 |