Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACTAGCTCCTGGAAGTAAGTTC[A/G]TTAGCTTTCCCCTCAACGTGCATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601062 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GUCD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GUCD1 - guanylyl cyclase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284251.1 | Intron | NP_001271180.1 | ||||
NM_001284252.1 | Intron | NP_001271181.1 | ||||
NM_001284253.1 | Intron | NP_001271182.1 | ||||
NM_001284254.1 | Intron | NP_001271183.1 | ||||
NM_001284255.1 | Intron | NP_001271184.1 | ||||
NM_001284256.1 | Intron | NP_001271185.1 | ||||
NM_001284257.1 | Intron | NP_001271186.1 | ||||
NM_031444.3 | Intron | NP_113632.2 | ||||
XM_005261761.1 | Intron | XP_005261818.1 |
SNRPD3 - small nuclear ribonucleoprotein D3 polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278656.1 | Intron | NP_001265585.1 | ||||
NM_004175.4 | Intron | NP_004166.1 |