Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCAGGAGCTGGGTGAGGCGGGG[C/T]AGCGCCCACAGGCTGGGCAGCAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612346 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GGT1 - gamma-glutamyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288833.1 | 650 | Intron | NP_001275762.1 | |||
NM_013421.2 | 650 | Intron | NP_038265.2 | |||
NM_013430.2 | 650 | Intron | NP_038347.2 |
LRRC75B - leucine rich repeat containing 75B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207644.2 | 650 | Silent Mutation | CTA,CTG | L,L 207 | NP_997527.2 | |
XM_005261600.2 | 650 | Intron | XP_005261657.1 |