Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACAGAGCTCAATAAATGCTCATT[C/T]GAATCTGGATAAGAGCAGTATCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604373 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHEK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHEK2 - checkpoint kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005735.1 | Intron | NP_001005735.1 | ||||
NM_001257387.1 | Intron | NP_001244316.1 | ||||
NM_007194.3 | Intron | NP_009125.1 | ||||
NM_145862.2 | Intron | NP_665861.1 | ||||
XM_006724114.3 | Intron | XP_006724177.2 | ||||
XM_006724116.2 | Intron | XP_006724179.2 | ||||
XM_011529839.2 | Intron | XP_011528141.1 | ||||
XM_011529840.2 | Intron | XP_011528142.1 | ||||
XM_011529841.1 | Intron | XP_011528143.1 | ||||
XM_011529842.2 | Intron | XP_011528144.1 | ||||
XM_011529843.1 | Intron | XP_011528145.1 | ||||
XM_011529844.2 | Intron | XP_011528146.1 | ||||
XM_011529845.2 | Intron | XP_011528147.1 | ||||
XM_017028560.1 | Intron | XP_016884049.1 | ||||
XM_017028561.1 | Intron | XP_016884050.1 |