Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAGGCAAGCTCTTTTGCTCACAG[A/G]GAGGGGTGCAGGTAGGAGTGCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613414 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL17REL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL17REL - interleukin 17 receptor E like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001694.2 | 3074 | UTR 3 | NP_001001694.2 | |||
XM_011530689.2 | 3074 | Intron | XP_011528991.1 | |||
XM_017028798.1 | 3074 | UTR 3 | XP_016884287.1 |