Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGAGGGAGGGGCTCAGCTTCCCC[C/G]GGTCACTGGGGCCAGGGAGACGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605908 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MLC1 - megalencephalic leukoencephalopathy with subcortical cysts 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015166.3 | 2334 | UTR 3 | NP_055981.1 | |||
NM_139202.2 | 2334 | UTR 3 | NP_631941.1 | |||
XM_011530678.1 | 2334 | UTR 3 | XP_011528980.1 | |||
XM_017028671.1 | 2334 | UTR 3 | XP_016884160.1 |
TTLL8 - tubulin tyrosine ligase like 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |