Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGACACAAGCGAGAGTGGTGCAGG[A/T]CTAACTCGCTTCCAGGTAAGGCCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300470 MIM: 300662 | ||||||||||||||||||||
Literature Links: |
MAGED2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAGED2 - MAGE family member D2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014599.5 | 223 | Silent Mutation | GGA,GGT | G,G 10 | NP_055414.2 | |
NM_177433.2 | 223 | Silent Mutation | GGA,GGT | G,G 10 | NP_803182.1 | |
NM_201222.2 | 223 | Silent Mutation | GGA,GGT | G,G 10 | NP_957516.1 |
SNORA11 - small nucleolar RNA, H/ACA box 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |