Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGGCTTTAGTTTAGAGCCTAAAG[A/G]GTTTGATAAACTTGTTGCCCTCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ARMCX5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARMCX5 - armadillo repeat containing, X-linked 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168478.1 | 1590 | Missense Mutation | GAG,GGG | E,G 311 | NP_001161950.1 | |
NM_001168479.1 | 1590 | Missense Mutation | GAG,GGG | E,G 311 | NP_001161951.1 | |
NM_001168480.1 | 1590 | Missense Mutation | GAG,GGG | E,G 311 | NP_001161952.1 | |
NM_001168482.1 | 1590 | Missense Mutation | GAG,GGG | E,G 311 | NP_001161954.1 | |
NM_001168485.1 | 1590 | Missense Mutation | GAG,GGG | E,G 311 | NP_001161957.1 | |
NM_022838.3 | 1590 | Missense Mutation | GAG,GGG | E,G 311 | NP_073749.2 |
ARMCX5-GPRASP2 - ARMCX5-GPRASP2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199818.1 | 1590 | Intron | NP_001186747.1 |