Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAAGCTGAGGCACACATTGGTT[A/G]AGTAGCTTAGCCAACCTCACTGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300921 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMCX5-GPRASP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARMCX5-GPRASP2 - ARMCX5-GPRASP2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199818.1 | Intron | NP_001186747.1 |
BHLHB9 - basic helix-loop-helix domain containing, class B, 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142524.1 | Intron | NP_001135996.1 | ||||
NM_001142525.1 | Intron | NP_001135997.1 | ||||
NM_001142526.1 | Intron | NP_001135998.1 | ||||
NM_001142527.1 | Intron | NP_001135999.1 | ||||
NM_001142528.1 | Intron | NP_001136000.1 | ||||
NM_001142529.1 | Intron | NP_001136001.1 | ||||
NM_001142530.1 | Intron | NP_001136002.1 | ||||
NM_030639.2 | Intron | NP_085142.1 |
GPRASP2 - G protein-coupled receptor associated sorting protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |