Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAGAAGAACGCTGGTAAGTACA[C/T]GAGGTGGGCATTGCAAGTGGCATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300689 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STARD8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STARD8 - StAR related lipid transfer domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142503.2 | Intron | NP_001135975.1 | ||||
NM_001142504.2 | Intron | NP_001135976.1 | ||||
NM_014725.4 | Intron | NP_055540.2 | ||||
XM_005262314.4 | Intron | XP_005262371.1 | ||||
XM_005262315.3 | Intron | XP_005262372.1 | ||||
XM_011531069.2 | Intron | XP_011529371.1 | ||||
XM_011531070.2 | Intron | XP_011529372.1 |