Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTAGTAAGTTACTCAGCTTCTGT[C/T]AGCCTTCATTGCCTCACCTGTGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 304040 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GJB1 - gap junction protein beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000166.5 | Intron | NP_000157.1 | ||||
NM_001097642.2 | Intron | NP_001091111.1 | ||||
XM_011530907.2 | Intron | XP_011529209.1 | ||||
XM_017029408.1 | Intron | XP_016884897.1 |