Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATTTAAAGAGCTTAGTTCTTTC[C/T]GCACTAAGACAGACATAAGTCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300282 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENOX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENOX2 - ecto-NOX disulfide-thiol exchanger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281736.1 | Intron | NP_001268665.1 | ||||
NM_006375.3 | Intron | NP_006366.2 | ||||
NM_182314.2 | Intron | NP_872114.1 | ||||
XM_005262354.3 | Intron | XP_005262411.1 | ||||
XM_011531244.2 | Intron | XP_011529546.1 | ||||
XM_011531245.2 | Intron | XP_011529547.1 | ||||
XM_011531247.2 | Intron | XP_011529549.1 | ||||
XM_011531249.2 | Intron | XP_011529551.1 | ||||
XM_011531251.2 | Intron | XP_011529553.1 | ||||
XM_017029226.1 | Intron | XP_016884715.1 | ||||
XM_017029227.1 | Intron | XP_016884716.1 | ||||
XM_017029228.1 | Intron | XP_016884717.1 | ||||
XM_017029229.1 | Intron | XP_016884718.1 |