Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATCAATGAGAGAATTCTTTCTG[C/T]ATATATGCAAATGTTCAGCGAGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 311030 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MCF2 - MCF.2 cell line derived transforming sequence | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099855.1 | Intron | NP_001093325.1 | ||||
NM_001171876.1 | Intron | NP_001165347.1 | ||||
NM_001171877.1 | Intron | NP_001165348.1 | ||||
NM_001171878.1 | Intron | NP_001165349.1 | ||||
NM_001171879.1 | Intron | NP_001165350.1 | ||||
NM_005369.4 | Intron | NP_005360.3 | ||||
XM_005262413.4 | Intron | XP_005262470.1 | ||||
XM_011531339.2 | Intron | XP_011529641.1 | ||||
XM_017029529.1 | Intron | XP_016885018.1 | ||||
XM_017029530.1 | Intron | XP_016885019.1 | ||||
XM_017029531.1 | Intron | XP_016885020.1 | ||||
XM_017029532.1 | Intron | XP_016885021.1 |