Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGAGTTCCCCTGAGTGTACTCA[A/C]AGTACTTTTGAGGGTTTTCCCCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300223 MIM: 300469 | ||||||||||||||||||||
Literature Links: |
MAGEC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAGEC1 - MAGE family member C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005462.4 | 1833 | Silent Mutation | CAA,CAC | Q,H 503 | NP_005453.2 | |
XM_011531418.2 | 1833 | Missense Mutation | CAA,CAC | Q,H 503 | XP_011529720.1 |
MAGEC3 - MAGE family member C3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |