Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAAGAACTGAAAAAGGAGGCAAA[G/T]AATGAAGCCAAAAATGATGCCCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 400005 | ||||||||||||||||||||
Literature Links: |
USP9Y PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USP9Y - ubiquitin specific peptidase 9, Y-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004654.3 | 1845 | Missense Mutation | AAG,AAT | K,N 300 | NP_004645.2 | |
XM_017030078.1 | 1845 | Missense Mutation | AAG,AAT | K,N 300 | XP_016885567.1 |