Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTTCGTCCTTGTGTAGTTTATG[A/G]TGGTGCTGATATTGGTCAGCAGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 400010 | ||||||||||||||||||||
Literature Links: |
DDX3Y PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX3Y - DEAD-box helicase 3, Y-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122665.3 | 1317 | Missense Mutation | GAT,GGT | D,G 300 | NP_001116137.1 | |
NM_001302552.2 | 1317 | Intron | NP_001289481.1 | |||
NM_001324195.1 | 1317 | Intron | NP_001311124.1 | |||
NM_004660.4 | 1317 | Intron | NP_004651.2 |