Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTGATCGTAGGAGAGGAGGATGC[C/T]GCTGTTGCAGCAGCAGCAGCTGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 490000 | ||||||||||||||||||||
Literature Links: |
ZFY PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZFY - zinc finger protein, Y-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145275.1 | 1187 | Intron | NP_001138747.1 | |||
NM_001145276.1 | 1187 | Silent Mutation | GCC,GCT | A,A 138 | NP_001138748.1 | |
NM_003411.3 | 1187 | Silent Mutation | GCC,GCT | A,A 329 | NP_003402.2 | |
XM_005262570.2 | 1187 | Silent Mutation | GCC,GCT | A,A 329 | XP_005262627.1 | |
XM_011531473.2 | 1187 | Silent Mutation | GCC,GCT | A,A 303 | XP_011529775.1 | |
XM_011531474.2 | 1187 | Silent Mutation | GCC,GCT | A,A 303 | XP_011529776.1 | |
XM_011531475.2 | 1187 | Intron | XP_011529777.1 | |||
XM_017030075.1 | 1187 | Silent Mutation | GCC,GCT | A,A 329 | XP_016885564.1 | |
XM_017030076.1 | 1187 | Intron | XP_016885565.1 | |||
XM_017030077.1 | 1187 | Intron | XP_016885566.1 |