Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTAAAGCCATAAGCTGCGATGG[A/G]ACCTAACCTCTGTGCCCCTTCAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610526 MIM: 616796 | ||||||||||||||||||||
Literature Links: |
NT5C1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NT5C1B - 5'-nucleotidase, cytosolic IB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002006.2 | 1853 | Missense Mutation | CCC,TCC | P,S 598 | NP_001002006.1 | |
NM_001199086.1 | 1853 | Missense Mutation | CCC,TCC | P,S 581 | NP_001186015.1 | |
NM_001199087.1 | 1853 | Missense Mutation | CCC,TCC | P,S 615 | NP_001186016.1 | |
NM_001199088.1 | 1853 | Missense Mutation | CCC,TCC | P,S 600 | NP_001186017.1 | |
NM_033253.3 | 1853 | Missense Mutation | CCC,TCC | P,S 538 | NP_150278.2 |
NT5C1B-RDH14 - NT5C1B-RDH14 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199103.1 | 1853 | Intron | NP_001186032.1 | |||
NM_001199104.1 | 1853 | Intron | NP_001186033.1 |
RDH14 - retinol dehydrogenase 14 (all-trans/9-cis/11-cis) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |