Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGTCATCTCAATGCCCATACCA[C/T]GGCTCTCATAAGGAGAGGAACTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 152425 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSL1 - acyl-CoA synthetase long-chain family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286708.1 | Intron | NP_001273637.1 | ||||
NM_001286710.1 | Intron | NP_001273639.1 | ||||
NM_001286711.1 | Intron | NP_001273640.1 | ||||
NM_001286712.1 | Intron | NP_001273641.1 | ||||
NM_001995.3 | Intron | NP_001986.2 | ||||
XM_005262828.1 | Intron | XP_005262885.1 | ||||
XM_005262829.1 | Intron | XP_005262886.1 | ||||
XM_005262831.1 | Intron | XP_005262888.1 | ||||
XM_011531742.1 | Intron | XP_011530044.1 | ||||
XM_017007887.1 | Intron | XP_016863376.1 | ||||
XM_017007888.1 | Intron | XP_016863377.1 | ||||
XM_017007889.1 | Intron | XP_016863378.1 |
SLED1 - proteoglycan 3 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |