Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAGGATGGGCTGCTGTTTACTTT[C/T]GGTGCTGGAAAACATGGGCAACTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608242 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HERC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HERC5 - HECT and RLD domain containing E3 ubiquitin protein ligase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016323.3 | 1467 | Silent Mutation | TTC,TTT | F,F 267 | NP_057407.2 | |
XM_011532022.2 | 1467 | Silent Mutation | TTC,TTT | F,F 418 | XP_011530324.2 |