Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAAAAGATGGGTGCCAGATATTG[C/T]GGGATCTGGCCAGCAAACCACAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615004 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRIT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRIT3 - leucine rich repeat, Ig-like and transmembrane domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198506.4 | Intron | NP_940908.3 | ||||
XM_017008167.1 | Intron | XP_016863656.1 | ||||
XM_017008168.1 | Intron | XP_016863657.1 |