Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATGAGAGAGAGATTCAGCCTACT[A/G]CTATTTTGTCCTTAGACCAGATCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602465 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPRY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPRY1 - sprouty RTK signaling antagonist 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258038.1 | 302 | Missense Mutation | ACT,GCT | T,A 37 | NP_001244967.1 | |
NM_001258039.1 | 302 | Missense Mutation | ACT,GCT | T,A 37 | NP_001244968.1 | |
NM_005841.2 | 302 | Missense Mutation | ACT,GCT | T,A 37 | NP_005832.1 | |
NM_199327.2 | 302 | Missense Mutation | ACT,GCT | T,A 37 | NP_955359.1 | |
XM_005262686.1 | 302 | Missense Mutation | ACT,GCT | T,A 37 | XP_005262743.1 | |
XM_017007635.1 | 302 | Missense Mutation | ACT,GCT | T,A 37 | XP_016863124.1 |