Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACCTTCTCTAAGGATTTTTCTCC[C/T]TCACTGTGTTCAGACATTGCAGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF150 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF150 - ring finger protein 150 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020724.1 | 6906 | Intron | NP_065775.1 | |||
XM_005263150.4 | 6906 | UTR 3 | XP_005263207.1 | |||
XM_011532147.2 | 6906 | UTR 3 | XP_011530449.1 | |||
XM_011532148.2 | 6906 | UTR 3 | XP_011530450.1 | |||
XM_017008475.1 | 6906 | UTR 3 | XP_016863964.1 | |||
XM_017008476.1 | 6906 | UTR 3 | XP_016863965.1 |