Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCCGAGAACGTGAAAAACAACTA[A/G]GAGCATCAAGAAGCACTATAGATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609550 | ||||||||||||||||||||
Literature Links: |
RNF150 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF150 - ring finger protein 150 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF330 - zinc finger protein 330 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292002.1 | 295 | UTR 5 | NP_001278931.1 | |||
NM_014487.5 | 295 | Missense Mutation | AGA,GGA | R,G 23 | NP_055302.1 | |
XM_017008033.1 | 295 | Missense Mutation | AGA,GGA | R,G 23 | XP_016863522.1 |