Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGCACTCCAAGGACCTCCTCTC[C/T]CACATGTGTTGTCAAATGACACCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609746 MIM: 600983 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP10 - Rho GTPase activating protein 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NR3C2 - nuclear receptor subfamily 3 group C member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000901.4 | 4824 | UTR 3 | NP_000892.2 | |||
NM_001166104.1 | 4824 | UTR 3 | NP_001159576.1 | |||
XM_011531975.1 | 4824 | UTR 3 | XP_011530277.1 | |||
XM_011531976.2 | 4824 | UTR 3 | XP_011530278.1 | |||
XM_011531977.2 | 4824 | UTR 3 | XP_011530279.1 | |||
XM_011531978.2 | 4824 | Intron | XP_011530280.1 | |||
XM_017008217.1 | 4824 | UTR 3 | XP_016863706.1 |