Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGATTTTAAGGCTTTTAATAACA[C/T]CTTGTTTCATAGCTATACATTTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605928 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARFIP1 - ADP ribosylation factor interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIGD4 - tigger transposable element derived 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145720.3 | 1783 | Missense Mutation | GAT,GGT | D,G 321 | NP_663772.1 | |
XM_005262807.4 | 1783 | Missense Mutation | GAT,GGT | D,G 321 | XP_005262864.1 |