Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAATTTTATTAATTCTGCCTTAG[A/C]GACATTTTCCAGTTTGCTCAGATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138247 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIA2 - glutamate ionotropic receptor AMPA type subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000826.3 | 135 | Intron | NP_000817.2 | |||
NM_001083619.1 | 135 | Intron | NP_001077088.1 | |||
NM_001083620.1 | 135 | Intron | NP_001077089.1 | |||
XM_017008113.1 | 135 | UTR 5 | XP_016863602.1 | |||
XM_017008114.1 | 135 | Intron | XP_016863603.1 | |||
XM_017008115.1 | 135 | Intron | XP_016863604.1 | |||
XM_017008116.1 | 135 | Intron | XP_016863605.1 | |||
XM_017008117.1 | 135 | UTR 5 | XP_016863606.1 |