Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATGGAAATAATTAATCAATTTT[C/G]CTTTCAATGAATTGTATACTGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607989 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPOCK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPOCK3 - sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040159.1 | 2737 | UTR 3 | NP_001035249.1 | |||
NM_001204352.1 | 2737 | UTR 3 | NP_001191281.1 | |||
NM_001204353.1 | 2737 | UTR 3 | NP_001191282.1 | |||
NM_001204354.1 | 2737 | UTR 3 | NP_001191283.1 | |||
NM_001204355.1 | 2737 | UTR 3 | NP_001191284.1 | |||
NM_001204356.1 | 2737 | UTR 3 | NP_001191285.1 | |||
NM_001204358.1 | 2737 | Intron | NP_001191287.1 | |||
NM_001251967.1 | 2737 | UTR 3 | NP_001238896.1 | |||
NM_016950.2 | 2737 | UTR 3 | NP_058646.2 | |||
XM_011532018.1 | 2737 | UTR 3 | XP_011530320.1 | |||
XM_017008257.1 | 2737 | UTR 3 | XP_016863746.1 | |||
XM_017008258.1 | 2737 | UTR 3 | XP_016863747.1 |