Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGAGATTTGGGTGGGGACACAG[C/T]CAAACCATATCAGTGGGTATGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601624 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCN2 - ficolin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004108.2 | Intron | NP_004099.2 | ||||
NM_015837.2 | Intron | NP_056652.1 | ||||
XM_006717015.3 | Intron | XP_006717078.1 | ||||
XM_011518392.2 | Intron | XP_011516694.1 |
LOC101448202 - uncharacterized LOC101448202 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |