Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGGTGGCAGCGAGACATGCACC[C/G]GGCCCGGAAGCTCCTCAGCCTCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605366 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OLFM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OLFM1 - olfactomedin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282611.1 | 502 | Intron | NP_001269540.1 | |||
NM_001282612.1 | 502 | Intron | NP_001269541.1 | |||
NM_006334.3 | 502 | Missense Mutation | CCG,CGG | P,R 13 | NP_006325.1 | |
NM_014279.4 | 502 | Missense Mutation | CCG,CGG | P,R 13 | NP_055094.1 |