Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTCTGGATGGTACTGATGTTAG[C/T]GTGGACTTGCCAGTAAGTAGGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPB41L5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPB41L5 - erythrocyte membrane protein band 4.1 like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184937.1 | 359 | Silent Mutation | AGC,AGT | S,S 56 | NP_001171866.1 | |
NM_020909.3 | 359 | Silent Mutation | AGC,AGT | S,S 56 | NP_065960.2 | |
XM_006712651.1 | 359 | Silent Mutation | AGC,AGT | S,S 56 | XP_006712714.1 | |
XM_006712652.1 | 359 | Silent Mutation | AGC,AGT | S,S 56 | XP_006712715.1 | |
XM_006712653.1 | 359 | UTR 5 | XP_006712716.1 | |||
XM_011511529.2 | 359 | Silent Mutation | AGC,AGT | S,S 56 | XP_011509831.1 | |
XM_017004567.1 | 359 | Silent Mutation | AGC,AGT | S,S 56 | XP_016860056.1 | |
XM_017004568.1 | 359 | UTR 5 | XP_016860057.1 |