Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTCTTTTCCTCCCTTTTTGTTC[C/T]AGTGCATTTAGTCAGGCCCTGTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165230 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLI2 - GLI family zinc finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005270.4 | Intron | NP_005261.2 | ||||
XM_006712422.2 | Intron | XP_006712485.1 | ||||
XM_011510969.2 | Intron | XP_011509271.2 | ||||
XM_011510970.2 | Intron | XP_011509272.1 | ||||
XM_011510971.2 | Intron | XP_011509273.1 | ||||
XM_011510972.2 | Intron | XP_011509274.2 | ||||
XM_011510973.2 | Intron | XP_011509275.1 | ||||
XM_011510974.2 | Intron | XP_011509276.1 | ||||
XM_017003818.1 | Intron | XP_016859307.1 |