Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCATTTAGGATTAACAGCGACAA[A/C]CTGTGAGTACGAGGAGCTGGGGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610519 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNTNAP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNTNAP5 - contactin associated protein like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130773.3 | 444 | Missense Mutation | AAC,ACC | N,T 27 | NP_570129.1 | |
XM_006712258.1 | 444 | Missense Mutation | AAC,ACC | N,T 27 | XP_006712321.1 | |
XM_017003316.1 | 444 | Missense Mutation | AAC,ACC | N,T 27 | XP_016858805.1 |