Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGAGGTGGACTTTCCAGCCTTG[A/G]GCACGACAGGATACTCCTTTCTGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609697 | ||||||||||||||||||||
Literature Links: |
SAP130 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SAP130 - Sin3A associated protein 130 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145928.1 | 2937 | Silent Mutation | NP_001139400.1 | |||
NM_024545.3 | 2937 | Silent Mutation | NP_078821.2 | |||
XM_005263767.3 | 2937 | Silent Mutation | XP_005263824.1 | |||
XM_005263768.3 | 2937 | Silent Mutation | XP_005263825.1 | |||
XM_005263769.3 | 2937 | Silent Mutation | XP_005263826.1 | |||
XM_005263770.3 | 2937 | Silent Mutation | XP_005263827.1 | |||
XM_006712748.3 | 2937 | Silent Mutation | XP_006712811.1 | |||
XM_006712749.3 | 2937 | Silent Mutation | XP_006712812.1 | |||
XM_017004903.1 | 2937 | Silent Mutation | XP_016860392.1 | |||
XM_017004904.1 | 2937 | Silent Mutation | XP_016860393.1 | |||
XM_017004905.1 | 2937 | Silent Mutation | XP_016860394.1 |