Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCCAAGGCTCAGGACCACCCCCA[C/T]TGCCTGTGAACACATTTGCATAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605216 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF4 - Rho guanine nucleotide exchange factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015320.3 | 3861 | Intron | NP_056135.2 | |||
NM_032995.2 | 3861 | Intron | NP_127462.1 |
FAM168B - family with sequence similarity 168 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009993.3 | 3861 | UTR 3 | NP_001009993.2 | |||
NM_001321743.1 | 3861 | UTR 3 | NP_001308672.1 | |||
NM_001321744.1 | 3861 | UTR 3 | NP_001308673.1 | |||
NM_001321745.1 | 3861 | UTR 3 | NP_001308674.1 | |||
NM_001321746.1 | 3861 | UTR 3 | NP_001308675.1 | |||
NM_001321747.1 | 3861 | UTR 3 | NP_001308676.1 | |||
NM_001321748.1 | 3861 | UTR 3 | NP_001308677.1 | |||
XM_011510594.2 | 3861 | UTR 3 | XP_011508896.1 | |||
XM_017003327.1 | 3861 | UTR 3 | XP_016858816.1 | |||
XM_017003328.1 | 3861 | UTR 3 | XP_016858817.1 |
POTEI - POTE ankyrin domain family member I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277406.1 | 3861 | Intron | NP_001264335.1 | |||
XM_011511670.2 | 3861 | Intron | XP_011509972.1 | |||
XM_017004732.1 | 3861 | Intron | XP_016860221.1 | |||
XM_017004733.1 | 3861 | Intron | XP_016860222.1 | |||
XM_017004734.1 | 3861 | Intron | XP_016860223.1 | |||
XM_017004735.1 | 3861 | Intron | XP_016860224.1 |