Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACAGCCAATGGATATTTATCAA[G/T]AATATAAAAATCTGTATCATACTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603084 | ||||||||||||||||||||
Literature Links: |
DARS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DARS - aspartyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293312.1 | 1358 | Missense Mutation | ATT,CTT | I,L 291 | NP_001280241.1 | |
NM_001349.3 | 1358 | Missense Mutation | ATT,CTT | I,L 391 | NP_001340.2 | |
XM_017003489.1 | 1358 | Missense Mutation | ATT,CTT | I,L 358 | XP_016858978.1 |