Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAGACTCATGGTAACACCAAAGC[A/G]ATTTTAAGAATGTGAATAGGAAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4782 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4782 - microRNA 4782 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC35F5 - solute carrier family 35 member F5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005263799.3 | 3361 | UTR 3 | XP_005263856.1 | |||
XM_011511921.2 | 3361 | UTR 3 | XP_011510223.1 | |||
XM_011511922.2 | 3361 | UTR 3 | XP_011510224.1 | |||
XM_011511923.2 | 3361 | UTR 3 | XP_011510225.1 | |||
XM_017005027.1 | 3361 | Intron | XP_016860516.1 | |||
XM_017005028.1 | 3361 | UTR 3 | XP_016860517.1 |